Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStrip™ Urinalysis Reagent Test Strips

Catalog No. p-3347649
Click to view available options
Detectable Analytes:
Glucose, Protein

Rapid screening tool detects up to four analytes on one test strip

  • Read strips visually or on instrumentation
  • Rapid qualitative results
  • Shelf life 18 months from date of manufacture
  • CLIA waived

Specifications

Clia Complexity Waived
Detectable Analytes Glucose, Protein
For Use With (Application) Urinalysis
Quantity 100 Tests
Test Time 1 min.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.