Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Germanium

Catalog No. 503099622 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
Catalog No. Quantity
50-309-9622 1 Ea.
50-309-9623 5 Ea.
2 options

Catalog No. 50-309-9622

Supplier: Sigma Aldrich GF588807391EA

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Germanium

Specifications

CAS 7440-56-4
Melting Point 937°C
Density 5.35 g/mL (at 25°C)
Boiling Point 2830°C
Molecular Formula Ge
Linear Formula Ge
Quantity 1 Ea.
MDL Number MFCD00085310
Synonym Germanium; GE004700
Molecular Weight (g/mol) 72.64
Percent Purity 100%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.