Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germanium(II) telluride, 99.999%

Catalog No. p-4824719
Click to view available options
Quantity:
1 g
5 g

CAS: 12025-39-7 | GeH2Te | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12025-39-7
Molecular Formula GeH2Te
Molecular Weight (g/mol) 202.25
MDL Number MFCD00135541
InChI Key SBMPISTWTKUJKL-UHFFFAOYSA-N
Synonym germanium telluride, germanium tel-luride, tellanylidenegermanylidene, germanium telluride trace metals basis
PubChem CID 16213264
IUPAC Name germanetellone
SMILES [GeH2]=[Te]

Specifications

Melting Point 725°C
Color Black
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.999%
Formula Weight 200.19
Density 6.14
Chemical Name or Material Germanium(II) telluride
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 234-706-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.