Learn More
Germanium(IV) chloride, 99.9999% (metals basis)
Description
It is used as a catalyst in the conversion of carbohydrates into 5-hydroxymethylfurfural in ionic liquids, as a reducing agent in combination with triphenylphosphine (TPP) for the reduction of alpha-bromo carboxylic acid derivatives, and in the production of pure germanium. It also finds use as an intermediate for several optical processes. It is one of the most important dopants in silica glass for optical fibers. It is employed in the preparation of germanium dioxide which is used for wide camera lens, microscopy, IR-transparent glasses/windows/lenses, and for the core of fiber-optic lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 83.1°C |
Melting Point | -49.5°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 500 g |
Linear Formula | GeCl4 |
UN Number | UN3264 |
Merck Index | 14,4410 |
IUPAC Name | germanium tetrachloride |
Formula Weight | 214.4 |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | GHS P Statement P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S. |
RTECSNumber | LY5225000 |
TSCA | TSCA |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.