Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germanium(IV) ethoxide, 97%

Catalog No. AA1467906
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 14165-55-0

It is a sol-gel intermediate for optical fibers and photosensitive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14165-55-0
MDL Number MFCD00015121
Synonym Germanium tetraethoxide; Tetraethoxygermane

Specifications

Melting Point -72°C
Color Colorless
Density 1.14 g/mL
Boiling Point 185.5°C
Flash Point 49°C (121°F)
Odor Alcohol-like
Refractive Index 1.407
Linear Formula Ge(OC2H5)4
Quantity 5 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Soluble in anhydrous alcohol and benzene; Hydrolyzes in water
Formula Weight 252.84
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Germanium(IV) ethoxide
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 238-007-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.