Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germanium, plasma standard solution, Specpure™ Ge 1000μg/mL

Catalog No. p-4824721
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA13841AD 50 mL
AA13841AE 100 mL
AA13841AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA13841AD Supplier Thermo Scientific Chemicals Supplier No. 013841AD
Only null left
Add to Cart

CAS: 7440-56-4 | Ge | 72.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-56-4 , 7664-39-3 , 7697-37-2 , 7732-18-5
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Hydrogen fluoride: 0.1%; Germanium: 0.1%; Water: 94.8%
Chemical Name or Material Germanium
Hazard Category H290-H315-H318
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
DOTInformation Hazard Class: 8, Packing Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.