Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Safety Tags

Catalog No. 17998149
Click to view available options
Message:
CAUTION
DANGER
DO NOT OPERATE
DO NOT START
DO NOT USE
Material:
Pulp-free Cardstock
RP Plastic
Length (Metric):
13 cm
14.6 cm
Style:
Standard back A
Standard back B

Provide immediate identification of the hazard by using the appropriate message for your tag

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145, ANSI Z535
Color Black, Yellow
Diameter (English) Grommet 0.37 in.
Material RP Plastic
Finish Matte
Message CAUTION
For Use With (Application) For indoor and outdoor conditions
Height (English) 0.01 in.
Height (Metric) 0.02cm
Length (Metric) 13 cm
Length (English) 5.12 in.
Thickness 15 mil (0.02 cm)
Style Standard back A
Width (English) 3.3 in.
Width (Metric) 8.38 cm
Dimensions (L x W) 5.12 x 3.3 in. (13 x 8.38 cm)
Diameter (Metric) Hole 0.93 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.