Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GET4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP141084

Catalog No. NBP141084


Only null left
Add to Cart

Description

Description

GET4 Polyclonal specifically detects GET4 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

GET4
Polyclonal
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
GET4
The immunogen for this product maps to a region between residue 277 and 327 of human conserved edge expressed protein using the numbering given in entry NP_057033.2 (GeneID 51608).
0.1 mL
Primary
Human
IgG
Western Blot, Immunoprecipitation
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
CEEchromosome 7 open reading frame 20, CGI-20, golgi to ER traffic protein 4 homolog (S. cerevisiae), TRC3535kDa
Rabbit
Affinity Purified
RUO
51608
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only