Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GF 109203X
GREENER_CHOICESDP

Catalog No. 074110 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-411-0 10 mg
07-411 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-411-0 Supplier R&D Systems™ Supplier No. 0741/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein kinase C inhibitor

Very potent and selective inhibitor of protein kinase C, selective for the α and β1 isoforms (IC50 values are 0.0084, 0.0180, 0.210, 0.132, and 5.8 μM for α, β1, δ, ε and ζ isoforms respectively). Selective over MLCK, PKG and PKA (IC50 values are 0.6, 4.6, and 33 μM respectively). Potent antagonist at the 5-HT3 receptor (Ki = 29.5 nM). Anti-inflammatory in vivo.

Chemical Identifiers

CAS 133052-90-1
Molecular Formula C25H24N4O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 412.49
Purity >98%
Molecular Weight (g/mol) 412.493
Chemical Name or Material GF 109203X
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.