Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Antibody (012), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289203AF700
Description
GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse samples. It is validated for Western Blot.Specifications
GFAP | |
Monoclonal | |
Alexa Fluor 700 | |
FLJ45472, GFAP astrocytes, glial fibrillary acidic protein | |
This GFAP Antibody (012) was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the human GFAP. | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
12 | |
50 mM Sodium Borate | |
Rabbit | |
50 kDa | |
0.1 mL | |
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
2670 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction