Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Antibody (012), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289203MFV610
Description
GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse samples. It is validated for Western BlotSpecifications
GFAP | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
012 | |
Western Blot | |
ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human GFAP. | |
0.1 mL | |
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
2670 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction