Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Antibody (GA-5), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 11 publications
$560.00
Specifications
Antigen | GFAP |
---|---|
Clone | GA-5 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 0.5 - 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.5 - 1 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 2 ug/mL, Immunohistochemistry-Paraffin 0.5 - 1 ug/mL, Immunohistochemistry-Frozen 0.5 - 1 ug/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
GFAP Monoclonal specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
GFAP | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
P14136 | |
2670 | |
GFAP isolated from pig spinal cord (Uniprot: P14136) | |
Primary | |
Store at 4C. | |
50 kDa |
GA-5 | |
Western Blot 0.5 - 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.5 - 1 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 2 ug/mL, Immunohistochemistry-Paraffin 0.5 - 1 ug/mL, Immunohistochemistry-Frozen 0.5 - 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Bovine, Chicken, Rabbit | |
FLJ45472, GFAP astrocytes, GFAP elisa, GFAP flow cytometry, glial fibrillary acidic protein | |
GFAP | |
IgG1 κ | |
Protein A or G purified | |
This monoclonal antibody recognizes a protein of ∼50kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title