Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Antibody, Novus Biologicals™
SDP

Catalog No. NB300141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300141 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300141 Supplier Novus Biologicals Supplier No. NB300141
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 66 publications

GFAP Polyclonal specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen GFAP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:5000, Simple Western 1:10000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation Supplied as serum with 5mM Sodium Azide
Gene Accession No. P14136, Q28115
Gene Alias FLJ45472, GFAP astrocytes, GFAP immunohistochemistry, GFAP mouse, GFAP rabbit, GFAP stain, glial fibrillary acidic protein
Gene Symbols GFAP
Host Species Rabbit
Immunogen This GFAP Antibody was developed against recombinant full length human GFAP isotype 1 expressed in and purified from E. coli.
Molecular Weight of Antigen 50 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Test Specificity Human, mouse, rat, chicken, bovine, porcine, rabbit, guinea pig, canine and feline. Predicted to work with most mammals.
Target Species Human, Mouse, Rat, Pig, Bovine, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.