Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11093480
Description
GFAP Polyclonal specifically detects GFAP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GFAP | |
Polyclonal | |
Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:1000 | |
Q28115 | |
GFAP | |
This GFAP Antibody was developed against recombinant and purified bovine GFAP. Accession # Q28115 | |
Unpurified | |
RUO | |
Primary | |
Specific for the 50 kDa GFAP protein. A lower band at 45 kDa is a proteolytic fragment derived from the GFAP molecule. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Neat whole antisera with No Preservative | |
FLJ45472, GFAP astrocytes, glial fibrillary acidic protein | |
Rabbit | |
50 kDa | |
0.1 mL | |
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
2670 | |
Human, Mouse, Rat, Bovine | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction