Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP (Glial Fibrillary Acidic Protein) Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1376R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1376R1 1 mL Prediluted Prediluted
MS1376P1 500 μL Concentrated 200μg/mL
MS1376P 1 mL Concentrated 200μg/mL
MS1376R7 7 mL Prediluted Prediluted
MS1376P0 100 μL Concentrated 200μg/mL
MS1376P1ABX 100 μL Concentrated 1mg/mL
MS1376PABX 200 μL Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1376R1 Supplier Epredia Supplier No. MS1376R1

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Glial Fibrillary Acidic Protein (GFAP) is specific to astrocytes (i.e., glial cells) and ependymal cells of the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS. Ab-6 stains astrocytes, glial cells, ependymal cells and their corresponding tumors. Many types of neural tumors such as neuroblastomas, Schwannomas, as well as extra-CNS tumors are not labeled.

Host Species: Mouse

Clone: ASTRO6

Isotype: IgG1

Species Reactivity: Human, Pig, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Glial Fibrillary Acidic Protein

Molecular Weight: 51-52 kDa

Positive Control: IMR-5 cells, Brain or astrocytoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

General References:

1. Trivino A et al. Vision Res, 1992, 32(9):1601-7.

2. Yachnis AT et al. Journal of Comparative Neurology, 1993, 334(3):356-69.

3. Debus E, et. al. Differentiation, 1983, 25(2):193-203.

Specifications

Antigen GFAP (Glial Fibrillary Acidic Protein)
Applications Immunofluorescence
Classification Monoclonal
Clone ASTRO6
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Glial Fibrillary Acidic Protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Chicken, Human, Pig, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.