Learn More
GFAP (Glial Fibrillary Acidic Protein) Rabbit Polyclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Monkey, Cow, Sheep, Guinea Pig, Chicken, Mouse, Rat and Hamster. Others not known.
Epitope: Not determined
Immunogen: GFAP isolated from cow spinal cord
Molecular Weight: 51-52 kDa
Positive Control: Brain or astrocytoma
Cellular Localization: Cytoplasmic Ab-4 stains astrocytes, glial cells, ependymal cells and their corresponding tumors. In the peripheral nervous system, Schwann cells, satellite cells, and enteric glial cells are stained. Weak staining of axons has been observed. Absorption studies show that this is caused by a cross-reaction with neurofilament protein. Ab-4 is particularly useful for distinguishing neoplasms of astrocytic origin from other neoplasms in the central nervous system by immunohistochemistry.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | GFAP (Glial Fibrillary Acidic Protein) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | GFAP isolated from cow spinal cord |
| Purification Method | Brain or astrocytoma |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
Frequently Asked Questions (FAQs)
Please note that only antibodies against neuronal proteins are specific to neuronal cells. We carry antibodies to GFAP, HuC/HuD, NMDA receptor, synapsin I as well as many other antibodies for neuronal proteins. A link to our primary antibody search tool can be found:
Neurobiology antibody search (https://www.thermofisher.com/us/en/home/life-science/antibodies/primary-antibodies/neurobiology-antibodies.html)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.