Learn More
GFAP Monoclonal Antibody (GA-5), Zeta
Mouse Monoclonal Antibody Cocktail
$497.76
Specifications
Antigen | GFAP |
---|---|
Clone | GA-5 |
Concentration | 100 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-221-5951
![]() |
ZETA CORPORATION
Z2255ML |
1 mL |
Each of 1 for $497.76
|
|
|||||
Description
A recommended positive control tissue for this product is Brain, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
GFAP Monoclonal antibody specifically detects GFAP in Human, Porcine samples. It is validated for Immunohistochemistry (Paraffin)Specifications
GFAP | |
100 μg/mL | |
Monoclonal | |
Liquid | |
RUO | |
tris with BSA, NP-40 and <0.1% sodium azide | |
6330404F12Rik; 65 kDa glutamic acid decarboxylase; AI836096; ALXDRD; Astrocyte or Intermediate Filament Protein; cb345; class III intermediate filament protein; etID36982.3; FLJ45472; GAD(65); Gad2; Gad-2; GAD65; GAD-65; GFAP; GFAP epsilon; gfap protein; gfapl; glial fibrillary acidic protein; Glial Fibrillary Acidic Protein (GFAP); glial fibrillary acidic protein alpha; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; glutamic acid decarboxylase 2; I79_011607; intermediate filament; intermediate filament protein; intermediate filament protein; GFAP; Unknown (protein for MGC:139638); wu:fb34h11; wu:fk42c12; xx:af506734; zgc:110485; zGFAP; intermediate filament protein; zrf-1 antigen | |
Native GFAP purified from pig spinal cord | |
Primary | |
2-8°C | |
GFAP |
GA-5 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Pig | |
P14136 | |
2670, 396562 | |
IgG1 κ | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.