Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Monoclonal Antibody (OTI1A7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF814532


Description
GFAP Monoclonal antibody specifically detects GFAP in Human samples. It is validated for ELISASpecifications
GFAP | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16552 | |
Mouse | |
Protein A/G | |
RUO | |
3605 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
ELISA | |
OTI1A7 | |
Unconjugated | |
IL17A | |
6330404F12Rik; 65 kDa glutamic acid decarboxylase; AI836096; ALXDRD; Astrocyte or Intermediate Filament Protein; cb345; class III intermediate filament protein; etID36982.3; FLJ45472; GAD(65); Gad2; Gad-2; GAD65; GAD-65; GFAP; GFAP epsilon; gfap protein; gfapl; glial fibrillary acidic protein; Glial Fibrillary Acidic Protein (GFAP); glial fibrillary acidic protein alpha; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; glutamic acid decarboxylase 2; I79_011607; intermediate filament; intermediate filament protein; intermediate filament protein; GFAP; Unknown (protein for MGC:139638); wu:fb34h11; wu:fk42c12; xx:af506734; zgc:110485; zGFAP; intermediate filament protein; zrf-1 antigen | |
Full length human recombinant protein of human IL-17A (NP_002181) produced in 293 cell. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction