Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Mouse anti-Human, Unconjugated, Clone: GFAP/2076, Abnova™

Catalog No. 89337506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-506 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-506 Supplier Abnova Corporation Supplier No. MAB21013
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GFAP Monoclonal antibody specifically detects GFAP in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen GFAP
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GFAP/2076
Conjugate Unconjugated
Description glial fibrillary acidic protein
Gene GFAP
Gene Alias FLJ45472
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 101-200 of human GFAP
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2670
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.