Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit, Clone: ASTRO/1974R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30787720UG
Description
GFAP Monoclonal specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
GFAP | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1 - 2 ug/million cells, Immunohistochemistry-Paraffin 1 - 2 ug/mL | |
ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
This GFAP Antibody (ASTRO/1974R) was developed against recombinant human full-length GFAP protein. | |
20 μg | |
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
2670 | |
Store at 4°C. | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
ASTRO/1974R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Chicken, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction