Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFER/ALR Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297019PE
Description
GFER/ALR Polyclonal antibody specifically detects GFER/ALR in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
GFER/ALR | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
ALRFAD-linked sulfhydryl oxidase ALR, Augmenter of liver regeneration, EC 1.8.3.2, ERV1 homolog, ERV1hepatopoietin protein, erv1-like growth factor, growth factor, augmenter of liver regeneration, growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration), Hepatopoietin, hERV1, HERV1hepatic regenerative stimulation substance, HPO, HPO1, HPO2, HSS, truncated augmenter of liver regeneration | |
Produced in rabbits immunized with purified, recombinant Human GFER/ALR (Accession#: NP_005253.3; Met1-Asp125) | |
0.1 mL | |
Cell Biology, Endocrinology, Signal Transduction | |
2671 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction