Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFER Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548256

Catalog No. PA548256


Only null left
Add to Cart

Description

Description

GFER Polyclonal Antibody for Western Blot, Flow

Isoform 1: FAD-dependent sulfhydryl oxidase that regenerates the redox-active disulfide bonds in CHCHD4/MIA40, a chaperone essential for disulfide bond formation and protein folding in the mitochondrial intermembrane space. The reduced form of CHCHD4/MIA40 forms a transient intermolecular disulfide bridge with GFER/ERV1, resulting in regeneration of the essential disulfide bonds in CHCHD4/MIA40, while GFER/ERV1 becomes re- oxidized by donating electrons to cytochrome c or molecular oxygen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GFER
Polyclonal
Unconjugated
Gfer
ALR; Augmenter of liver regeneration; ERV1; ERV1 homolog; erv1-like growth factor; FAD-linked sulfhydryl oxidase ALR; Gfer; growth factor, augmenter of liver regeneration; growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration); growth factor, erv1 homolog; growth factor, erv1-like (augmenter of liver regeneration); hepatic regenerative stimulation substance; Hepatopoietin; hepatopoietin protein; HERV1; HPO; HPO1; HPO2; HSS; MNCb-0663
Rabbit
Ammonium sulfate precipitation
RUO
11692, 2671
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P55789, P56213
Gfer
KLH conjugated synthetic peptide between 173-202 amino acids from the C-terminal region of human GFER.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.