Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFER Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566596 Supplier Invitrogen™ Supplier No. PA566596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RFHGGNLFFLP GGARSEMMDD LATD Highest antigen sequence identity to the following orthologs - mouse 64%, rat 60%.

Isoform 1: FAD-dependent sulfhydryl oxidase that regenerates the redox-active disulfide bonds in CHCHD4/MIA40, a chaperone essential for disulfide bond formation and protein folding in the mitochondrial intermembrane space. The reduced form of CHCHD4/MIA40 forms a transient intermolecular disulfide bridge with GFER/ERV1, resulting in regeneration of the essential disulfide bonds in CHCHD4/MIA40, while GFER/ERV1 becomes re- oxidized by donating electrons to cytochrome c or molecular oxygen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFER
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gfer
Gene Accession No. P55789
Gene Alias ALR; Augmenter of liver regeneration; ERV1; ERV1 homolog; erv1-like growth factor; FAD-linked sulfhydryl oxidase ALR; Gfer; growth factor, augmenter of liver regeneration; growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration); growth factor, erv1 homolog; growth factor, erv1-like (augmenter of liver regeneration); hepatic regenerative stimulation substance; Hepatopoietin; hepatopoietin protein; HERV1; HPO; HPO1; HPO2; HSS; MNCb-0663
Gene Symbols Gfer
Host Species Rabbit
Immunogen Recombinant Human GFER. Recombinant protein control fragment (Product #RP-107251).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2671
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.