Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFI-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | GFI-1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 6-8ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227173SS
![]() |
Novus Biologicals
NBP227173SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227173
![]() |
Novus Biologicals
NBP227173 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
GFI-1 Polyclonal specifically detects GFI-1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GFI-1 | |
Western Blot 6-8ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine, Chicken | |
GFI-1, growth factor independence-1, growth factor independent 1, growth factor independent 1 transcription repressor, Growth factor independent protein 1, SCN2, Zinc finger protein 163, zinc finger protein Gfi-1, ZNF163FLJ94509 | |
GFI1 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chicken and cow. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
2672 | |
A portion of amino acid 280-330 of human Zinc finger protein Gfi-1 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title