Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GFI-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$200.50 - $561.50
Specifications
Antigen | GFI-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
GFI-1 Polyclonal specifically detects GFI-1 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
GFI-1 | |
Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
GFI1, GFI-1, growth factor independence-1, growth factor independent 1, growth factor independent 1 transcription repressor, Growth factor independent protein 1, Pal-1, SCN2, Zinc finger protein 163, zinc finger protein Gfi-1, ZNF163, ZNF163FLJ94509 | |
GFI1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Q99684 | |
2672 | |
E. coli-derived recombinant human GFI-1 Pro2-Leu250 Accession # Q99684 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human GFI-1 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title