Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFI1 Monoclonal Antibody (OTI6H7), TrueMAB™, OriGene
SDP

Catalog No. 501680747
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-0747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-0747 Supplier OriGene Supplier No. TA805546
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Gfi-1 is a nuclear zinc finger (ZF) transcriptional repressor that plays an important role in hematopoiesis and inner ear development, and has been implicated in lymphomagenesis. The Gfi1 protein expression is regulated by the ubiquitin-proteasome system. It is reported that ubiquitin ligase Triad1 interacts with the DNA-binding domain of Gfi-1. Gfi-1 represses transcription by directly binding to the consensus DNA sequence in the promoters of its target genes. Deficiency of Gfi-1 causes neutropenia and an accumulation of granulomonocytic precursor cells that is reminiscent of a myelodysplastic syndrome in mice.

Specifications

Antigen GFI1
Applications Western Blot
Classification Monoclonal
Clone OTI6H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GFI1
Gene Accession No. Q99684
Gene Alias GFI-1, GFI1A, SCN2, ZNF163
Gene Symbols GFI1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-180 of human GFI1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2672
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.