Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFI1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540328 Supplier Invitrogen™ Supplier No. PA540328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MPRSFLVKSK MAHTYHQPRV QEDEPLWPPA LTPVPRDQAP SNSPVLSTLF Sequence homology: Cow: 83%; Dog: 100%; Goat: 83%; Horse: 100%; Human: 100%; Mouse: 83%; Pig: 93%; Rat: 82%.

GFI1b (zinc finger protein GFI-1b) is an essential proto-oncogenic transcriptional regulator necessary for development and differentiation of erythroid and megakaryocytic lineages. It is also a component of a RCOR-GFI-KDM1A-HDAC complex that suppresses, via histone deacetylase (HDAC) recruitment, a number of genes implicated in multilineage blood cell development and controls hematopoietic differentiation. GFI1b also acts as a transcriptional repressor or activator depending on both promoter and cell type context; represses promoter activity of SOCS1 and SOCS3 and thus, may regulate cytokine signaling pathways. It also cooperates with GATA1 to repress target gene transcription, such as the apoptosis regulator BCL2L1; GFI1B silencing in leukemic cell lines markedly increase apoptosis rate. Mutations affecting the gene are associated with Bleeding disorder, platelet-type 17 (BDPLT17).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFI1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GFI1B
Gene Accession No. Q5VTD9
Gene Alias BDPLT17; GFI1B; Gfi-1B; growth factor independent 1B; growth factor independent 1B (potential regulator of CDKN1A, translocated in CML); growth factor independent 1B transcription repressor; growth factor independent 1B transcriptional repressor; growth factor independent 1B-like protein; growth factor independent protein 1B; growth factor-independent 1B; Potential regulator of CDKN1A translocated in CML; Zinc finger protein Gfi-1b; ZNF163B
Gene Symbols GFI1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GFI1B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8328
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.