Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFI1B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GFI1B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GFI1B Polyclonal specifically detects GFI1B in Mouse samples. It is validated for Western Blot.Specifications
GFI1B | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
growth factor independent 1B transcription repressor, Growth factor independent protein 1B, Potential regulator of CDKN1A translocated in CML, translocated inCML), zinc finger protein Gfi-1b | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human GFI1B (NP_001128503.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS with 50% glycerol, pH7.3. | |
8328 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title