Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GFM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111521
Description
Immunogen sequence: NKLDRMGSNP ARALQQMRSK LNHNAAFMQI PMGLEGNFKG IVDLIEERAI YFDGDFGQIV RYGEIPAELR AAATDHRQEL IECVANSDEQ LGEMFLE.
Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. Mitochondrial translation is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. This gene encodes one of the mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known.
Specifications
| GFM1 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96RP9 | |
| GFM1 | |
| Recombinant Protein corresonding to Human GFM1. Recombinant protein control fragment (Product #RP-105185). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| GFM1 | |
| AW545374; COXPD1; D3Wsu133e; Efg; EF-G; Efg1; EFGM; EF-Gmt; EF-Gmt {ECO:0000255; EGF1; Elongation factor G 1, mitochondrial; elongation factor G 1, mitochondrial {ECO:0000255; elongation factor G, mitochondrial; elongation factor G, mitochondrial {ECO:0000255; Elongation factor G1; elongation factor G1 {ECO:0000255; G elongation factor 1; G elongation factor mitochondrial 1; G elongation factor, mitochondrial 1; G translation elongation factor, mitochondrial; GFM; Gfm1; HAMAP-Rule:MF_03061}; hEFG1; mEF-G 1; mEF-G 1 {ECO:0000255; mitochondrial; mitochondrial elongation factor G; mitochondrial elongation factor G1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85476 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction