Learn More
GFM1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | GFM1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-9672
![]() |
Bethyl Laboratories, Inc
A305151AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and X. laevis
Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. Mitochondrial translation is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintece of mitochondrial DNA. This gene encodes one of the mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known.Specifications
GFM1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
COXPD1, EFG, EFG1, EFGM, EGF1, GFM, hEFG1, EF-Gmt, G translation elongation factor, mitochondrial, elongation factor G 1, mitochondrial, elongation factor G, mitochondrial, elongation factor G1, mEF-G 1, mitochondrial elongation factor G, mitochondrial elongation factor G1 | |
Between 525 and 575 | |
Primary | |
4° C | |
GFM1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8K0D5, Q96RP9 | |
28030, 85476 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.