Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFM1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982815
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2815 Supplier Bioss Supplier No. BS13338R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. Mitochondrial translation is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. This gene encodes one of the mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known.

Specifications

Antigen GFM1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GFM1
Gene Accession No. Q96RP9
Gene Alias AW545374; BOS_1164; COXPD1; D3Wsu133e; Efg; EF-G; EFG1; EFGM; EF-Gmt; EF-Gmt {ECO:0000255; EGF1; Elongation factor G 1, mitochondrial; elongation factor G 1, mitochondrial {ECO:0000255; Elongation factor G, mitochondrial; elongation factor G, mitochondrial {ECO:0000255; elongation factor G, mitochondrial-like; Elongation factor G1; elongation factor G1 {ECO:0000255; G elongation factor 1; G elongation factor mitochondrial 1; G elongation factor, mitochondrial 1; G translation elongation factor, mitochondrial; Gfm; Gfm1; HAMAP-Rule:MF_03061}; hEFG1; LOC100055837; LOC100345180; mEF-G 1; mEF-G 1 {ECO:0000255; mitochondrial; mitochondrial elongation factor G; mitochondrial elongation factor G1
Gene Symbols GFM1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GFM1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85476
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.