Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFM2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13339R

Description
GFM2 is involved in mitochondrial translation, which is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. In Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. This gene encodes one of mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known. Alternative splicing results in at least three transcript variants encoding distinct isoforms.Specifications
GFM2 | |
Polyclonal | |
Unconjugated | |
Gfm2 | |
6530419G12Rik; A930009M04Rik; EFG2; EF-G2mt; EF-G2mt {ECO:0000255; Elongation factor G 2, mitochondrial; elongation factor G 2, mitochondrial {ECO:0000255; Elongation factor G2; G elongation factor; G elongation factor mitochondrial 2; G elongation factor, mitochondrial 2; Gfm2; HAMAP-Rule:MF_03059}; hEFG2; mEF-G 2; mEF-G 2 {ECO:0000255; mitochondrial elongation factor G2; mitochondrial ribosome recycling factor 2; MRRF2; MST027; MSTP027; RGD1309854; Ribosome-releasing factor 2, mitochondrial; ribosome-releasing factor 2, mitochondrial {ECO:0000255; RRF2; RRF2mt; RRF2mt {ECO:0000255 | |
Rabbit | |
Protein A | |
RUO | |
84340 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q969S9 | |
Gfm2 | |
KLH conjugated synthetic peptide derived from human GFM2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction