Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFOD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531269
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531269 Supplier Invitrogen™ Supplier No. PA531269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (94%), Rat (94%), Chicken (85%), Rhesus Monkey (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GFOD2, or glucose-fructose oxidoreductase domain-containing protein 2, is part of the small GFOD gene family in humans, which consists of only two members: GFOD1 and GFOD2. The gene encodes a protein that belongs to the Gfo/Idh/MocA family of oxidoreductases, which are known for their diverse substrate range and utilization of NAD or NADP as redox cofactors. Despite the presence of an enzyme-defining domain, the exact enzymatic activity and biological function of GFOD2 remain unclear. Research suggests that these proteins may not exhibit significant tissue distribution or function, indicating a possible pseudoenzyme role.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFOD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene GFOD2
Gene Accession No. Q3B7J2
Gene Alias 5730466C23Rik; GFOD2; glucose-fructose oxidoreductase domain containing 2; glucose-fructose oxidoreductase domain-containing protein 2; RGD1311198; UNQ9430/PRO34691
Gene Symbols GFOD2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GFOD2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81577
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.