Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFOD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559360 Supplier Invitrogen™ Supplier No. PA559360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVSMAASFED GLYMQSVVDA IKRSSRSGEW EAVEVLTEEP DTNQNLCEAL QRNNL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

GFOD2, or glucose-fructose oxidoreductase domain-containing protein 2, is part of the small GFOD gene family in humans, which consists of only two members: GFOD1 and GFOD2. The gene encodes a protein that belongs to the Gfo/Idh/MocA family of oxidoreductases, which are known for their diverse substrate range and utilization of NAD or NADP as redox cofactors. Despite the presence of an enzyme-defining domain, the exact enzymatic activity and biological function of GFOD2 remain unclear. Research suggests that these proteins may not exhibit significant tissue distribution or function, indicating a possible pseudoenzyme role.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFOD2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GFOD2
Gene Accession No. Q3B7J2
Gene Alias 5730466C23Rik; GFOD2; glucose-fructose oxidoreductase domain containing 2; glucose-fructose oxidoreductase domain-containing protein 2; RGD1311198; UNQ9430/PRO34691
Gene Symbols GFOD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GFOD2. Recombinant protein control fragment (Product #RP-98574).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81577
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.