Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB42401V100UG
This item is not returnable.
View return policy
Description
GFP Monoclonal specifically detects GFP in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
GFP | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P42212 | |
GFP | |
E. coli-derived recombinant GFPuv Ser2-Lys238 Accession # P42212 | |
100 μg | |
Primary | |
Multi-species | |
Purified |
Flow Cytometry | |
454505 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects recombinant GFPuv and eGFP in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction