Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody (1025527), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB42402MFV610
Description
GFP Monoclonal antibody specifically detects GFP in Broad Species Reactivity samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ImmunocytochemistrySpecifications
GFP | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, Immunocytochemistry | |
1025527 | |
Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
E. coli-derived recombinant GFP, aa 2-238 | |
0.1 mL | |
Epitope Tags | |
Broad Species Reactivity | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction