Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody (208), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290178AF750
Description
GFP Monoclonal antibody specifically detects GFP in Nonspecies specific samples. It is validated for Western Blot, ELISA.Specifications
GFP | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Non-species specific | |
Purified |
Western Blot, ELISA | |
208 | |
Western Blot, ELISA | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Aequorea victoria GFP (AAB65663; Ser2-Lys238). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction