Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody, Alkaline Phosphatase, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB6001502
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.Specifications
GFP | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.1% Sodium Azide | |
GFP | |
The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. (Uniprot: P42212) | |
0.1 mg | |
Epitope Tags | |
No reaction was observed against Human, Mouse and Rat Serum Proteins. | |
Store at 4°C. Do not freeze. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot, Immunoassay, KnockDown | |
1 mg/ml | |
Western Blot 1:500-1:2500, Simple Western 1:10, ELISA 1:2000-1:8000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Dot Blot, Knockout Validated, Knockdown Validated | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Jellyfish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction