Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600308B
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockdown Validated.Specifications
GFP | |
Polyclonal | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockdown Validated | |
anti-gfp (green fluorescent protein) pab, Enhanced Green Fluorescent Protein, GFP immunofluorescence, GFP immunoprecipitation, GFP mouse, GFP staining, GFP western blot, Green Fluorescent Protein, green fluorescent protein (gfp) | |
Rabbit | |
Affinity Purified | |
Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
No reaction was observed against Human, Mouse or Rat serum proteins. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Immunohistochemistry (Free Floating), KnockDown | |
Biotin | |
PBS with 0.05% Sodium Azide | |
GFP | |
The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. | |
0.1 mL | |
Primary | |
Non-species specific, C. elegans, Plant | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction