Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody, DyLight 488, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP169969
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay, Knockout Validated, Knockdown Validated.Specifications
GFP | |
Polyclonal | |
DyLight 488 | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
GFP | |
The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. (Uniprot: P42212) | |
0.1 mg | |
Primary | |
Reconstitute with 100 ul deionized water (or equivalent) | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot, Immunoassay, KnockDown | |
LYOPH | |
Western Blot 1:10000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence >1:5000, Dot Blot, Fluorophore-linked immunosorbent assay 1:100 - 1:2000, Knockout Validated, Knockdown Validated | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
Goat | |
Affinity Purified | |
RUO | |
No reaction was observed against Human, Mouse or Rat serum proteins. | |
Jellyfish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction