Learn More
Description
Specifications
Specifications
| Antigen | GFP |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1.1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:10000, Flow Cytometry 1:10 - 1:1000, ELISA 1:10000-1:30000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Proximity Ligation Assay, Electron Microscopy 1:10 - 1:500, Immunohistochemistry Free-Floating 1:10 - 1:500, Knockout Validated, Knockdown Validated |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | P42212 |
| Gene Alias | Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) |
| Gene Symbols | GFP |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
