Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody, Novus Biologicals™

Chicken Polyclonal Antibody has been used in 38 publications
Supplier: Novus Biologicals NB10016140.02ML
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.Specifications
| GFP | |
| Polyclonal | |
| Unconjugated | |
| 10mM PBS (0.9% isotonic, w/v, pH 7.2) and 50% Glycerol with 0.02% Sodium Azide | |
| GFP | |
| Chickens were immunized with purified recombinant green fluorescent protein (GFP) emulsified in adjuvant. After multiple injections, eggs were collected from The hens, and IgY fractions were prepared from The yolks. Purified antibody preparations were Then mixed with glycerol 1:1(v/v) (toprevent freezing at -20 degrees C), augmented with thimerosal, and then filter-sterilized. | |
| 0.02 mL | |
| Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
| GFP | |
| Store at -20C in the dark. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 10.0 mg/mL | |
| Western Blot 1:2500-1:5000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Jellyfish |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction