Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600308PE
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Flow Cytometry, Knockdown Validated.Specifications
GFP | |
Polyclonal | |
Flow Cytometry, KnockDown Validated | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. | |
0.1 mL | |
Primary | |
Non-species specific, C. elegans, Plant | |
IgG |
Flow Cytometry, KnockDown | |
PE | |
PBS with 0.05% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
No reaction was observed against Human, Mouse or Rat serum proteins. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction