Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB42401RSP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB42401RSP 25 μg
MAB42401RP 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB42401RSP Supplier R&D Systems Supplier No. MAB42401RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GFP Monoclonal specifically detects GFP in Multi-Species samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen GFP
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 454505R
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P42212
Gene Alias eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp)
Host Species Mouse
Immunogen E.coli-derived recombinant GFPuv Ser2-Lys238 Accession # P42212
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Stains eGFP transfectants but not irrelevant transfectants in flow cytometry. Detects recombinant GFPuv in Western blots. For optimal Western blot results, we recommend R&D Systems Goat Anti-GFP Antigen Affinity-purified Polyclonal Antibody (Catalog # AF4240 ) or Novus Biologicals Mouse Anti-GFP Monoclonal Antibody (Catalog # NBP2-2211 ).
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.