Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Monoclonal Antibody (A11-C11), Huabio

Catalog No. 502544485
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4485 Supplier HUABIO RESEARCH INC Supplier No. M12101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Stable for 12 months from date of receipt.

GFP Monoclonal antibody specifically detects GFP in Cnidarian samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation

Specifications

Antigen GFP
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone A11-C11
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP
Host Species Mouse
Immunogen Recombinant full length protein of Aequorea victoria GFP.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Cnidarian
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.