Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP, Mouse anti-All, Clone: 2B6, Millipore Sigma™

Catalog No. MABC1689100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1689100 100 μg
MABC168925 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1689100 Supplier MilliporeSigma Supplier No. MABC1689100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GFP, clone 2B6, Cat. No. MABC1689, is a mouse monoclonal antibody that detects GFP-conjugated to most proteins and has been tested for use in Immunocytochemistry and Western Blotting.

Green fluorescent protein (UniProt: P42212) is encoded by the GFP gene in Aequorea victoria (Jellyfish). Green fluorescence protein (GFP) is a 27 kDa protein from jellyfish that serves as an energy-transfer acceptor. It transduces the blue chemiluminescence of the protein aequorin into green fluorescent light by energy transfer and it fluoresces in vivo upon receiving energy from the Ca2+-activated photoprotein aequorin. GFP has an absorption max at 395 nm and the emission peaks at 507 to 510 nm. However, the exact value of emission max depends on the environment of the chromophore. GFP is an important tool in cell biology research that displays stability under fixation conditions and is suitable for a variety of applications, such as tracing cell lineage tracer, reporter of gene expression, or exploring protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFP
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2B6
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected GFP in HEK293T cells stabely transfected with GFP-tagged Aquaporin 4.
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Green fluorescent protein
Gene Symbols GFP
Host Species Mouse
Immunogen His-tagged recombinant jellyfish Green fluorescent protein.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary & Control Antibodies
Primary or Secondary Primary
Test Specificity Clone 2B6 is a mouse monoclonal antibody that detects GFP-conjugated to most proteins.
Target Species All species
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.