Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody (4B10B2), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222111PCP
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Flow Cytometry.Specifications
| GFP | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Non-species specific | |
| Purified |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4B10B2 | |
| Flow Cytometry | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Purified recombinant fragment of GFP expressed in E. coli. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction