Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4240P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4240P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4240P Supplier R&D Systems Supplier No. IC4240P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

GFP Polyclonal specifically detects GFP in Multi-Species samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen GFP
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P42212
Gene Alias eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Goat
Immunogen E. coli-derived recombinant GFPuv Ser2-Lys238 Accession # P42212
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects GFP in direct ELISAs and Western blots.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.