Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Rabbit anti-Jellyfish, Clone: 208, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | GFP |
---|---|
Clone | 208 |
Dilution | Western Blot 1:1000-1:5000, ELISA 1:10000-1:20000, Immunoprecipitation 1-4 ul/mg of lysate |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
GFP | |
Western Blot 1:1000-1:5000, ELISA 1:10000-1:20000, Immunoprecipitation 1-4 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
Recombinant Aequorea victoria GFP protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
208 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Jellyfish | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title