Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Rabbit anti-Nonspecies specific, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00
Specifications
Antigen | GFP |
---|---|
Dilution | Western Blot 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:2000 - 1:5000 |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GFP | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Epitope Tags | |
50% PBS and 50% Glycerol | |
This GFP antibody was developed against full length recombinant GFP. | |
Primary |
Western Blot 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:2000 - 1:5000 | |
Polyclonal | |
Purified | |
RUO | |
Non-species specific | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
IgG | |
Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title